Skip to Content
Merck
All Photos(1)

Documents

SAB4700808

Sigma-Aldrich

Monoclonal Anti-CD49d antibody produced in mouse

clone 9F10

Synonym(s):

ITGA4, Integrin alpha-4, VLA-4 alpha, VLA4 alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9F10, monoclonal

form

buffered aqueous solution

species reactivity

cat, dog, sheep, human, horse, bovine, nonhuman primates

concentration

1 mg/mL

technique(s)

flow cytometry: 6 μg/mL

accession no.

NM_000885

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ITGA4(3676)

Related Categories

General description

The mouse monoclonal antibody 9F10 recognizes CD49d (alpha 4 integrin), a 145-180 kDa type I transmembrane glycoprotein expressed on B and T cells, monocytes, eosinophils, basophils, NK cells, and dendritic cells, but not platelets.

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service