Skip to Content
Merck
All Photos(3)

Documents

SAB2701915

Sigma-Aldrich

Anti-col2a1a antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

col2a1, col2a1a, coll2a1, fb38c06

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

zebrafish

technique(s)

immunohistochemistry: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

zebrafish ... col2a1a(562496)

Immunogen

Recombinant protein containing a sequence corresponding to a region within amino acids 100 and 188 of col2a1a according to NP_571367.1

Application

Suggested starting dilutions are as follows: IHC-Wm: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anita Senk et al.
Scientific reports, 11(1), 19520-19520 (2021-10-02)
Although well investigated, the importance of collagen fibers in supporting angiogenesis is not well understood. In this study, we demonstrate that extracellular collagen fibers provide guidance cues for endothelial cell migration during regenerative angiogenesis in the caudal zebrafish fin. Inhibition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service