Skip to Content
Merck
All Photos(3)

Key Documents

SAB1402600

Sigma-Aldrich

Monoclonal Anti-RIOK2, (C-terminal) antibody produced in mouse

clone 1B10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ11159

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RIOK2(55781)

Immunogen

RIOK2 (AAH00953.1, 453 a.a. ~ 551 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
IALSSLNREFRPFRDEENVGAMNQYRTRTLSITSSGSAVSCSTIPPELVKQKVKRQLTKQQKSAVRRRLQKGEANIFTKQRRENMQNIKSSLEAASFWG

Biochem/physiol Actions

Serine/threonine-protein kinase RIO2 (RIOK2) is a non-ribosomal factor required for normal ribosomal RNA biogenesis and cell cycle progression. It is necessary for processing of 20S pre-rRNA to the 18S rRNA in 40S ribosomal subunit synthesis. RIOK2 is also a ribosomal assembly factor that prevents premature translation initiation on the small (40S) subunit. It is needed for proper mitotic progression, regulates metaphase-anaphase transition and is a substrate of Plk1 (polo-like kinase1) which is phosphorylated by Plk1 at the Ser-335, Ser-380, and Ser-548 residues.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorylation of Right Open Reading Frame 2 (Rio2) Protein Kinase by Polo-like Kinase 1 Regulates Mitotic Progression*
Ting Liu
The Journal of Biological Chemistry, 21, 286(42)-286(42) (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service