Skip to Content
Merck
All Photos(3)

Key Documents

SAB1402233

Sigma-Aldrich

Monoclonal Anti-HSPA4 antibody produced in mouse

clone 3A11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

APG-2, HS24/P52, MGC131852, RY, hsp70, hsp70RY

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~42.39 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPA4(3308)

Immunogen

HSPA4 (AAH02526, 1 a.a. ~ 148 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSVVGIDLGFQSCYVAVARAGGIETIANEYSDRCTPACISFGPKNRSIGAAAKSQVISNAKNTVQGFKRFHGRAFSDPFVEAEKSNLAYDIVQLPTGLTGIKVTYMEEEQNGPVDGQGDNPGPQAAEQGTDTAVPSDSDKKLPEMDID

Biochem/physiol Actions

Hspa4 (heat shock protein family A (Hsp70) member 4) is an important ATP-dependent cytosolic chaperone along with Hsp90. Hspa4 aids in the folding of several proteins and helps in protecting against cellular stresses such as rise in temperature. Hspa4 maintains the function of the mitotic centrosome to coordinates bipolar mitotic spindle assembly. Hspa4 might compensate the loss of parkin function in mice by protecting the cells against proteolytic and mitochondrial stress. Therefore, it can serve as an important therapeutic agent for parkinson disease. Upregulation of HSPA4 is observed in parkin gene knockout mice. Hspa4 is known to be associated with tumor progression and offers resistance against many therapeutic agents. The gene is upregulated in many different types of cancer.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacological or Genetic Activation of Hsp70 Protects against Loss of Parkin Function.
Zhang CW
Neuro-Degenerative Diseases, 16(5-6), 304-316 (2016)
Heat Shock Proteins Promote Cancer: It's a Protection Racket.
Calderwood SK and Gong J
Trends in Biochemical Sciences, 41(4), 311-323 (2016)
HSP70 regulates the function of mitotic centrosomes.
Fang CT
Cellular and Molecular Life Sciences, 73(20), 3949-3960 (2016)
Induction of Hsp70 in tumor cells treated with inhibitors of the Hsp90 activity: A predictive marker and promising target for radiosensitization.
Kudryavtsev VA, et.al.
PLoS ONE, 12(3), 1-25 (2017)
Huihui Ji et al.
PloS one, 12(3), e0172335-e0172335 (2017-03-03)
Aberrant DNA methylation has been observed in the patients with Alzheimer's disease (AD), a common neurodegenerative disorder in the elderly. OPRD1 encodes the delta opioid receptor, a member of the opioid family of G-protein-coupled receptors. In the current study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service