Skip to Content
Merck
All Photos(1)

Documents

SAB1401539

Sigma-Aldrich

Anti-CCR9 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CDw199, GPR-9-6, GPR28

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCR9(10803)

General description

The protein encoded by this gene is a member of the beta chemokine receptor family. It is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptors are key regulators of the thymocytes migration and maturation in normal and inflammation conditions. The specific ligand of this receptor is CCL25. It has been found that this gene is differentially expressed by T lymphocytes of small intestine and colon, suggested a role in the thymocytes recruitment and development that may permit functional specialization of immune responses in different segment of the gastrointestinal tract. This gene is mapped to the chemokine receptor gene cluster region. Two alternatively spliced transcript variants have been described. (provided by RefSeq)

Immunogen

CCR9 (NP_112477.1, 1 a.a. ~ 369 a.a) full-length human protein.

Sequence
MTPTDFTSPIPNMADDYGSESTSSMEDYVNFNFTDFYCEKNNVRQFASHFLPPLYWLVFIVGALGNSLVILVYWYCTRVKTMTDMFLLNLAIADLLFLVTLPFWAIAAADQWKFQTFMCKVVNSMYKMNFYSCVLLIMCISVDRYIAIAQAMRAHTWREKRLLYSKMVCFTIWVLAAALCIPEILYSQIKEESGIAICTMVYPSDESTKLKSAVLTLKVILGFFLPFVVMACCYTIIIHTLIQAKKSSKHKALKVTITVLTVFVLSQFPYNCILLVQTIDAYAMFISNCAVSTNIDICFQVTQTIAFFHSCLNPVLYVFVGERFRRDLVKTLKNLGCISQAQWVSFTRREGSLKLSSMLLETTSGALSL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service