Skip to Content
Merck
All Photos(3)

Key Documents

SAB1400439

Sigma-Aldrich

Anti-ARHGAP26 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-GRAF, Anti-KIAA0621, Anti-OPHN1L, Anti-OPHN1L1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Related Categories

Immunogen

ARHGAP26 (AAH68555.1, 1 a.a. ~ 759 a.a) full-length human protein.

Sequence
MGLPALEFSDCCLDSPHFRETLKSHEAELDKTNKFIKELIKDGKSLISALKNLSSAKRKFADSLNEFKFQCIGDAETDDEMCIARSLQEFATVLRNLEDERIRMIENASEVLITPLEKFRKEQIGAAKEAKKKYDKETEKYCGILEKHLNLSSKKKESQLQEADSQVDLVRQHFYEVSLEYVFKVQEVQERKMFEFVEPLLAFLQGLFTFYHHGYELAKDFGDFKTQLTISIQNTRNRFEGTRSEVESLMKKMKENPLEHKTISPYTMEGYLYVQEKRHFGTSWVKHYCTYQRDSKQITMVPFDQKSGGKGGEDESVILKSCTRRKTDSIEKRFCFDVEAVDRPGVITMQALSEEDRRLWMEAMDGREPVYNSNKDSQSEGTAQLDSIGFSIIRKCIHAVETRGINEQGLYRIVGVNSRVQKLLSVLMDPKTASETETDICAEWEIKTITSALKTYLRMLPGPLMMYQFQRSFIKAAKLENQESRVSEIHSLVHRLPEKNRQMLQLLMNHLANVANNHKQNLMTVANLGVVFGPTLLRPQEETVAAIMDIKFQNIVIEILIENHEKIFNTVPDMPLTNAQLHLSRKKSSDSKPPSCSERPLTLFHTVQSTEKQEQRNSIINSSLESVSSNPNSILNSSSSLQPNMNSSDPDLAVVKPTRPNSLPPNPSPTSPLSPSWPMFSAPSSPMPTSSTSSDSSPVSTPFRKAKALYACKAEHDSELSFTAGTVFDNVHPSQEPGWLEGTLNGKTGLIPENYVEFL

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mami Matsuda et al.
The Journal of general virology, 95(Pt 12), 2658-2667 (2014-08-07)
Previous studies have shown that hepatitis C virus (HCV) enters human hepatic cells through interaction with a series of cellular receptors, followed by clathrin-mediated, pH-dependent endocytosis. Here, we investigated the mechanisms of HCV entry into multiple HCV-permissive human hepatocyte-derived cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service