Skip to Content
Merck
All Photos(1)

Documents

PZ0155

Sigma-Aldrich

PF-03716556

≥98% (HPLC)

Synonym(s):

PF-03716556, [N-(2-Hydroxyethyl)-N,2-dimethyl-8-{[(4R)-5-methyl-3,4-dihydro-2H-chromen-4-yl]amino}imidazo[1,2-a]pyridine-6-carboxamide]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N4O3
CAS Number:
Molecular Weight:
394.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

CN(CCO)C(=O)c1cc(N[C@@H]2CCOc3cccc(C)c23)c4nc(C)cn4c1

InChI

1S/C22H26N4O3/c1-14-5-4-6-19-20(14)17(7-10-29-19)24-18-11-16(22(28)25(3)8-9-27)13-26-12-15(2)23-21(18)26/h4-6,11-13,17,24,27H,7-10H2,1-3H3/t17-/m1/s1

InChI key

YBHKBMJREUZHOV-QGZVFWFLSA-N

Biochem/physiol Actions

potent, selective and reversible acid pump antagonist (H, K-ATPase)

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to potassium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service