Skip to Content
Merck
All Photos(3)

Documents

PLA0128

Sigma-Aldrich

Rabbit anti-Phospho RNA Polymerase II (S2) Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

220kDa, DNA-directed RNA polymerase II largest subunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, POLR2, POLRA, RNA polymerase II 220 kd subunit, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1, polymerase (RNA) II (DNA directed) polypeptide A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:200- 1:1,000
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000-1:10,000

accession no.

NP_000928.1

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... POLR2A(5430)

Immunogen

Immunogen for PLA0128 was a phosphorylated synthetic peptide, which represented a portion of human RNA Polymerase II (GeneID 5430) around serine 2 of the C-terminal repeat YSPTSPS.

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

The RNA polymerase II subunit B1 (RPB1) is the largest subunit of the RNA polymerase II core complex. RNA polymerase II holoenzyme functions to transcribe DNA into messenger RNA in eukaryotes. RPB1 forms the DNA binding domain of the polymerase that forms a cleft that grabs the incoming DNA template to translocate the holoenzyme across the RNA-DNA hybrid.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service