Skip to Content
Merck
All Photos(1)

Key Documents

P9638

Sigma-Aldrich

L-α-Phosphatidylinositol 4-monophosphate sodium salt

≥98%

Synonym(s):

1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol 4-phosphate) sodium-potassium salt, Diphosphoinositide sodium salt, PIP, PtdInsP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

Assay

≥98%

form

powder

solubility

H2O: soluble

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Phosphorylated phospholipid membrane component, precursor of inositol phosphates. Formed in brain membranes from phosphatidylinositol by phosphatidylinositol 4-kinase.

Features and Benefits

This compound is featured on the Phosphoinositide Kinases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A O'Brian et al.
FEBS letters, 214(2), 339-342 (1987-04-20)
Protein kinase C(PKC) is a Ca2+- and phospholipid-dependent protein kinase which can be activated by diacylglycerol, a product of polyphosphoinositide hydrolysis. In this report, we show that the polyphosphoinositides L-alpha-phosphatidylinositol 4-monophosphate (PI 4P) and L-alpha-phosphatidylinositol 4,5-diphosphate (PI 4.5DP) can serve
L Raptis et al.
Journal of virology, 63(2), 753-758 (1989-02-01)
Polyomavirus middle tumor antigen (mT) was expressed in a line of mouse NIH 3T3 cells under control of the dexamethasone-regulatable mouse mammary tumor virus promotor. Contrary to rat F111 cells which were rendered anchorage independent by mT expression alone (L.
Siavash Fazel Darbandi et al.
Cell reports, 31(2), 107495-107495 (2020-04-16)
Tbr1 is a high-confidence autism spectrum disorder (ASD) gene encoding a transcription factor with distinct pre- and postnatal functions. Postnatally, Tbr1 conditional knockout (CKO) mutants and constitutive heterozygotes have immature dendritic spines and reduced synaptic density. Tbr1 regulates expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service