Key Documents
GN760180
Greiner CELLSTAR® serological pipette
volume 25 mL, 2 / 10 mL graduations, red nozzle
Synonym(s):
pipette, serological pipette, Greiner serological pipettes
About This Item
UNSPSC Code:
41121500
Pricing and availability is not currently available.
sterility
sterile
packaging
pack of 200 ea
manufacturer/tradename
Greiner 760180
availability
available only in China
General description
Greiner pipettes are precision extruded from the highest quality, optical-grade polystyrene. All volumes are identified by the standard international color code convention. Accuracy is maintained by high surface polish, both internally and externally, to reduce meniscus formation and ensure complete liquid expulsion with minimum carry-over.
Precision is aided by fine-line graduation printing providing accuracy of better than ±2%.
Features and Benefits
  • 1 mL, 2 mL and 5 mL sizes incorporate drawn tips while larger sizes feature molded tips to provide maximum accuracy
  • Packed individually, then double packed for extra protection
  • Designed to fit all standard pipetting devices
  • Cotton plugged for safety
  • Sterilized by γ-irradiation
  • DNase, RNase & human DNA free
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)