Skip to Content
Merck
All Photos(1)

Key Documents

GERPN3680

Alkphos Direct Labelling Reagents

Cytiva RPN3680

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106000
NACRES:
NA.31

packaging

pkg of 1 kit

manufacturer/tradename

Cytiva RPN3680

shipped in

wet ice

storage temp.

2-8°C

General description

AlkPhos Direct Labeling and Detection Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase. CDP-Star or ECF detection can be used

Each system includes the following reagents, sufficient for 25 or 50 labeling reactions and detection of 2500 cm2 or 5000 cm2 membrane (depending on product ordered): labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, CDP-Star or ECF detection reagents.

Features and Benefits

  • DNA and RNA probes are labelled during a 30-minute reaction.
  • Hybridization stringency can be controlled by temperature as well as salt concentration.
  • Rapid detection, 30-minute from completion of hybridization to application of detection reagent.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

AlkPhos Direct
This product is protected by patent numbers US 5,939,257 and EP 770678 owned by Cytiva Bio-Sciences Corp. and equivalent patents and patent applications in other countries.
ECF is a trademark of Cytiva

Pictograms

Environment

Signal Word

Danger

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service