Skip to Content
Merck
All Photos(1)

Key Documents

G5297

Sigma-Aldrich

Glp-Asn-Pro-d-Tyr-d-Trp-NH2

≥98% (HPLC), solid

Synonym(s):

JAK4D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H40N8O8
CAS Number:
Molecular Weight:
688.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: >2 mg/mL

storage temp.

−20°C

SMILES string

NC(=O)C[C@H](NC(=O)[C@@H]1CCC(=O)N1)C(=O)N2CCC[C@H]2C(=O)N[C@H](Cc3ccc(O)cc3)C(=O)N[C@H](Cc4c[nH]c5ccccc45)C(N)=O

InChI

1S/C34H40N8O8/c35-28(44)16-26(41-31(47)23-11-12-29(45)38-23)34(50)42-13-3-6-27(42)33(49)40-25(14-18-7-9-20(43)10-8-18)32(48)39-24(30(36)46)15-19-17-37-22-5-2-1-4-21(19)22/h1-2,4-5,7-10,17,23-27,37,43H,3,6,11-16H2,(H2,35,44)(H2,36,46)(H,38,45)(H,39,48)(H,40,49)(H,41,47)/t23-,24+,25+,26-,27-/m0/s1

InChI key

SZQSOKVXBMULDL-VQHLWIOESA-N

Gene Information

Amino Acid Sequence

Glp-Asn-Pro-Tyr-Trp-NH2

Biochem/physiol Actions

The synthetic peptide Glp-Asn-Pro-d-Tyr-d-Trp-NH2 mimics and amplifies central actions of TRH in rat without releasing TSH. Glp-Asn-Pro-d-Tyr-d-Trp-NH2 has dual pharmacological activities: it inhibits TRH-DE and binds to central TRH receptors with nM affinity in rat hippocampus and cortex but not pituitary or heterologous cells expressing TRH receptor 1 (TRHR1) or TRH receptor 2 (TRHR2). The peptide can be a potent tool for probing the role of TRH actions in the CNS and a platform for development of novel TRH-based therapeutics for neuro diseases.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service