Skip to Content
Merck
All Photos(1)

Key Documents

F6800

Sigma-Aldrich

Fenoldopam monohydrobromide

≥98%

Synonym(s):

6-Chloro-2,3,4,5-tetrahydro-1-(4-hydroxyphenyl)-1H-3-benzazepine-7,8-diol monohydrobromide, SKF-82526

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClNO3 · HBr
CAS Number:
Molecular Weight:
386.67
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98%

form

solid

storage condition

protect from light
under inert gas

color

off-white

solubility

DMSO: >12 mg/mL
H2O: 4.75 mg/mL

storage temp.

2-8°C

SMILES string

ClC1=C(O)C(O)=CC2=C1CCNCC2C3=CC=C(O)C=C3.[H]Br

InChI

1S/C16H16ClNO3.BrH/c17-15-11-5-6-18-8-13(9-1-3-10(19)4-2-9)12(11)7-14(20)16(15)21;/h1-4,7,13,18-21H,5-6,8H2;1H

InChI key

DSGOSRLTVBPLCU-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Fenoldopam is a selective dopamine agonist that is being considered for the parenteral treatment of systemic hypertension. In both an oral and parenteral form, fenoldopam causes peripheral vasodilation by stimulating dopamine-1 adrenergic receptors. Intravenous fenoldopam may provide advantages over sodium nitroprusside because it can induce both a diuresis and natriuresis, is not light sensitive, and is not associated with cyanide toxicity. There is no evidence for rebound hypertension after discontinuation of fenoldopam infusion.

Features and Benefits

This compound was developed by Hospira. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A de Vries et al.
Journal of cardiovascular pharmacology, 34(2), 191-198 (1999-08-13)
The natriuretic response to a dopamine 1-like receptor agonist is blunted in spontaneously hypertensive rats (SHRs). Whether the renal vasodilator response to D1-like receptor stimulation in SHRs is defective also is unclear. To determine whether the renal hemodynamic response to
M Quevedo et al.
General pharmacology, 32(1), 123-125 (1999-01-15)
1. Fenoldopam mesylate, a benzazepine derivative, is a D1 receptor agonist that lowers blood pressure through vasodilation of renal, mesenteric, coronary and cerebral vascular beds. 2. Experiments were performed in rats, and mean carotid blood pressure and heart rate were
R A Hahn et al.
The Journal of pharmacology and experimental therapeutics, 223(2), 305-313 (1982-11-01)
SK&F 82526 (6-chloro-7,8-dihydroxy-1-(p-hydroxyphenyl)-2,3,4,5-tetrahydro-(1H)-3-benzazepine) was evaluated for its peripheral cardiovascular activity, effect on renal and central dopamine receptors and mechanism of action. Comparisons were made with dopamine. In anesthetized dogs, SK&F 82526 i.v., produced dose-related dilation of the renal vasculature, apparently
T Hussain et al.
The American journal of physiology, 272(3 Pt 2), F339-F346 (1997-03-01)
The dopamine DA1 receptor transduces its signal via adenylyl cyclase and phospholipase C in the renal proximal tubule, which has been suggested to be defective at the level of receptor-G protein coupling in spontaneously hypertensive rats (SHR). We prepared basolateral
U Venkatakrishnan et al.
Clinical and experimental hypertension (New York, N.Y. : 1993), 22(3), 309-324 (2000-05-10)
Dopamine and dopamine-1 receptor agonists produce diuresis and natriuresis by causing changes in renal hemodynamics and by the activation of dopamine-1 receptors located within the various regions of the nephron. Nitric oxide plays an important role in the maintenance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service