Key Documents
EPM1190-0000
Eppendorf® New Brunswick Innova® Platform Shaker
2000, size 1.9 cm (3/4 in.) , orbit diameter, AC/DC input 120 V, 60 Hz (US)
Synonym(s):
laboratory shakers, orbital shakers
About This Item
UNSPSC Code:
12352200
feature
2000
CSA and CE compliant
benchtop model
platform sold separately
manufacturer/tradename
Eppendorf® M1190-0000x (US)
size
1.9 cm (3/4 in.) , orbit diameter
General description
platform shaker; benchtop model
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)