EP5810746000
Eppendorf® Adapter
holds 20x 1,2-5 mL tubes/ D = 11 mm, for rotor A-4-81, S-4x500, pack of 2 ea
Select a Size
€213.00
Select a Size
About This Item
€213.00
Recommended Products
feature
holds 20x 1,2-5 mL tubes/ D = 11 mm, for rotor A-4-81, S-4x500
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5810746000
technique(s)
centrifugation: suitable
compatibility
for use with 500 mL rectangular buckets
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service