Documents
EP5391000036
Eppendorf(R) HeatSealer
S100
About This Item
UNSPSC Code:
41102400
NACRES:
NC.21
AC/DC input
230 V, 50/60 Hz, UK plug
feature
CE compliant
CSA approved
fixed Sealing, temperature
seals 96 and 384 well plates
manufacturer/tradename
Eppendorf® 5391000036 (GB)
General description
The Eppendorf HeatSealer Family offers you reliable and effective Sealing, of a variety of plates. You can choose from a more compact (S100) and a more flexible (S200) instrument according to your Sealing, needs.
Features and Benefits
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
Legal Information
Eppendorf is a registered trademark of Eppendorf AG