Skip to Content
Merck
All Photos(1)

Key Documents

E9906

Sigma-Aldrich

Anti-EDEM2 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ER degradation enhancer, mannosidase alpha-like 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

human, rat (predicted), mouse

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using mouse 3T3 cells
western blot: 0.5-1 μg/mL using whole extracts of HEK-293T cells expressing recombinant human EDEM2

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EDEM2(55741)
mouse ... Edem2(108687)
rat ... Edem2(296304)

General description

ER degradation-enhancing α-mannosidase-like 2 is an enzyme encoded by the EDEM2 gene in humans. EDEM2 is localized to the endoplasmic reticulum (ER) mainly as a soluble glycoprotein. It is an enzyme encoded by the EDEM2 gene in humans. It is one of the ER-stress-induced members of the glycosyl hydrolase 47 family.

Immunogen

synthetic peptide corresponding to amino acids 22-38 of human EDEM2, conjugated to KLH. The corresponding sequence differs by one amino acid in mouse and 2 amino acids in rat EDEM2.

Application

Anti-EDEM2 (N-terminal) antibody produced in rabbit is suitable for indirect immunofluorescence at a concentration of 5-10μg/mL using mouse 3T3 cells and western blotting at a concentration of 0.5-1μg/mL using whole extracts of HEK-293T cells expressing recombinant human EDEM2.

Biochem/physiol Actions

Overexpression of ER degradation-enhancing alphamannosidase-like protein 2 (EDEM2) accelerates ER associated degradation (ERAD) by promoting the release of terminally misfolded glycoproteins from the calnexin cycle, without affecting the rate of degradation of nonglycosylated polypeptides or the maturation of model secretory proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of EDEM in the release of misfolded glycoproteins from the calnexin cycle
Molinari M, et al.
Science, 299(5611), 1397-1400 (2003)
Steven W Mast et al.
Glycobiology, 15(4), 421-436 (2004-11-13)
In the endoplasmic reticulum (ER), misfolded proteins are retrotranslocated to the cytosol and degraded by the proteasome in a process known as ER-associated degradation (ERAD). Early in this pathway, a proposed lumenal ER lectin, EDEM, recognizes misfolded glycoproteins in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service