Skip to Content
Merck
All Photos(1)

Key Documents

D1564

Sigma-Aldrich

Anti-DR6 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Death Receptor 6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 68 kDa

species reactivity

human

technique(s)

microarray: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

General description

DR6 is a type I transmembrane receptor and possesses four extracellular cysteine-rich motifs and a cytoplasmic death domain. It is expressed in most human tissues and its expression is found to be increased in heart, brain, placenta, pancreas, thymus, lymph node and several non-lymphoid cancer cell lines.

Immunogen

synthetic peptide corresponding to amino acids 42-56 of the N-terminal region of human DR6 precursor.

Physical form

Solution in 0.01 M phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Pan et al.
FEBS letters, 431(3), 351-356 (1998-08-26)
Tumor nectosis factor (TNF) receptors are key players in inflammation and immune regulation. A new member of this family, termed death receptor-6 (DR6), has been identified. Like other death receptors, DR6 is a type I transmembrane receptor, possesses four extracellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service