Skip to Content
Merck
All Photos(1)

Documents

CLS3789

Corning® 96 Well Solid Polystyrene Microplate

round bottom, white polystyrene, bag of 25, non-sterile, lid: no

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

material

round bottom
white bottom
white polystyrene

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 25

manufacturer/tradename

Corning 3789A

size

96 wells

well volume

330 μL

working volume

75-200 μL

color

white

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm2
  • Without lids, lids supplied separately
  • White microplates enhance luminescent signals and have low background luminescence and fluorescence
  • Designed to reduce well-to-well crosstalk
  • Total volume is 330μL; recommended working volume of 75 to 200 μL
  • Conforms to standard microplate footprint and dimensions

Note: Total volume is 330μL due to round bottom wells

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service