Skip to Content
Merck
All Photos(1)

Documents

CHO-K1.Cl-0238

15042911, hamster ovary, Polygonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CHO-K1.Cl-0238, 15042911, from hamster ovary

biological source

hamster ovary

description

CHO-K1, non-human primate FcγRIIa, Fc gamma receptor 3A, Macaca mulatta (Rhesus macaque)

growth mode

Adherent

morphology

Polygonal

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Stably transfected with a pcDNA3.1/Hygro(+) based vector with a cDNA insert that had been modified to enable expression of Rhesus macaque FcγRIIIa as a GPI linked receptor

Cell Line Description

Clone of CHO-K1 cells expressing Rhesus macque (Macaca mulatta) FcγRIIIa anchored to glycosyl phosphatidylinositol. (Predicted from DQ423379 NCBI sequence)

Culture Medium

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6. Seeding at 2-5 x 10,000 cells/cm2 using 0.05% trypsin/EDTA; culture conditions 5% CO2/95%air; 37oC

Other Notes

Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Citation

If use of this culture results in a scientific publication, it should be cited in the publication as: CHO-K1.Cl-0238 (ECACC 15042911)

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service