Skip to Content
Merck
All Photos(1)

Documents

KARPAS-929

06072606, human bone marrow, round to polygonal cells growing in clusters in suspension

Synonym(s):

KARPAS 929, KARPAS, 929, KARPAS929

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

KARPAS-929, from human bone marrow, 06072606

biological source

human bone marrow

growth mode

Suspension

karyotype

Hyperdiploid with complex aberrant chromosomes

morphology

Round to polygonal cells growing in clusters in suspension

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Related Categories

Cell Line Origin

Human multiple myeloma

Cell Line Description

Established in 1984 from the bone marrow of a 67-year old woman diagnosed with myeloma who survived for one year after diagnosis.

DNA Profile

Not specified

Culture Medium

RPMI 1640 + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Doubling time approximately 30 hours. Cells grow singly in suspension. Maintain cultures between 5-20 x 105 cells/ml; 5% CO2; 37 °C. Thaw ampoule and perform a cell count, spin out the DMSO and resuspend in an appropriate volume of media to give a cell concentration of 5 x 105 cells/ml. At subculture perform a cell count and maintain cultures between 5-20 x 105 cells/ml, this tends to equate to a 1:3 split every 2-3 days.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service