Skip to Content
Merck
All Photos(1)

Key Documents

C8091

Sigma-Aldrich

α-Chloralose

BioXtra, ≥98%

Synonym(s):

1,2-O-(2,2,2-Trichloroethylidene)-α-D-glucofuranose, Anhydro-D-glucochloral, Chloralose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11Cl3O6
CAS Number:
Molecular Weight:
309.53
Beilstein:
85418
EC Number:
MDL number:
UNSPSC Code:
12352201

product line

BioXtra

Assay

≥98%

impurities

<0.001% Phosphorus (P)
<0.1% Insoluble matter

ign. residue

<0.1%

solubility

ethanol: 0.1 M, clear, colorless

anion traces

sulfate (SO42-): <0.05%

cation traces

Al: <0.0005%
Ca: <0.002%
Cu: <0.0005%
Fe: <0.0005%
K: <0.005%
Mg: <0.0005%
NH4+: <0.05%
Na: <0.005%
Pb: <0.001%
Zn: <0.0005%

SMILES string

OC[C@@H](O)[C@H]1O[C@@H]2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl

InChI

1S/C8H11Cl3O6/c9-8(10,11)7-16-5-3(14)4(2(13)1-12)15-6(5)17-7/h2-7,12-14H,1H2/t2-,3+,4-,5-,6-,7-/m1/s1

InChI key

OJYGBLRPYBAHRT-IPQSZEQASA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service