Skip to Content
Merck
All Photos(1)

Key Documents

C7866

Sigma-Aldrich

CGP-53353

solid

Synonym(s):

5,6-Bis[(4-fluorophenyl)amino]-2H-isoindole-1,3-dione, DAPH 2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13F2N3O2
CAS Number:
Molecular Weight:
365.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Pricing and availability is not currently available.

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

solid

color

orange

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

Fc1ccc(Nc2cc3C(=O)NC(=O)c3cc2Nc4ccc(F)cc4)cc1

InChI

1S/C20H13F2N3O2/c21-11-1-5-13(6-2-11)23-17-9-15-16(20(27)25-19(15)26)10-18(17)24-14-7-3-12(22)4-8-14/h1-10,23-24H,(H,25,26,27)

InChI key

RONQPWQYDRPRGG-UHFFFAOYSA-N

Gene Information

human ... EGFR(1956)

Biochem/physiol Actions

Selective PKCβII inhibitor. IC50 - 410 nM for PKCβII vs 3.8 μM for PKCβI. Inhibits nPKCs and aPKCs only above 22 μM and other kinases such as protein kinase A, casein kinase I and II, c-src kinase, c-lyn kinase, c-fgr kinase, and v-abl kinase at an IC50 of greater than 50 μM.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Need A Sample COA?

    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service