Skip to Content
Merck
All Photos(1)

Key Documents

B7883

Sigma-Aldrich

Biotin methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18N2O3S
CAS Number:
Molecular Weight:
258.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

mp

~166.5 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(=O)OC)[C@@]1([H])NC(=O)N2

InChI

1S/C11H18N2O3S/c1-16-9(14)5-3-2-4-8-10-7(6-17-8)12-11(15)13-10/h7-8,10H,2-6H2,1H3,(H2,12,13,15)/t7-,8-,10-/m0/s1

InChI key

BHEWJAXNLVWPSC-NRPADANISA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Takeuchi et al.
Analytical chemistry, 72(11), 2418-2422 (2000-06-17)
Synthetic biotin-binding polymers were prepared by molecular imprinting. Methacrylic acid (MAA) was copolymerized with ethylene glycol dimethacrylate in the presence of biotin methyl ester (B-Me) in chloroform. Hydrogen-bonding-based complexation of B-Me with MAA generates the binding sites complementary to B-Me
Dolores Santa María et al.
The Journal of organic chemistry, 76(16), 6780-6788 (2011-07-12)
Four hosts (7-10) containing 2,6-bisamidopyridine- and 2,5-bisamidopyrrole-bearing pyridyl or 1,8-naphthyridyl groups have been prepared and their structures studied by a combination of multinuclear NMR spectroscopy and X-ray crystallography. Their behavior in molecular recognition of urea derivatives, including (+)-biotin methyl ester

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service