Skip to Content
Merck
All Photos(1)

Documents

B5383

Sigma-Aldrich

(+)-Biotin 4-nitrophenyl ester

Synonym(s):

4-Nitrophenyl (+)-biotinate, d-Biotin p-nitrophenyl ester, BNP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O5S
CAS Number:
Molecular Weight:
365.40
Beilstein:
5795805
MDL number:
UNSPSC Code:
12352000

mp

163-165 °C (lit.)

storage temp.

−20°C

SMILES string

[O-][N+](=O)c1ccc(OC(=O)CCCC[C@@H]2SCC3NC(=O)NC23)cc1

InChI

1S/C16H19N3O5S/c20-14(24-11-7-5-10(6-8-11)19(22)23)4-2-1-3-13-15-12(9-25-13)17-16(21)18-15/h5-8,12-13,15H,1-4,9H2,(H2,17,18,21)/t12-,13-,15-/m0/s1

InChI key

YUDNXDTXQPYKCA-YDHLFZDLSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Piffeteau et al.
Biochimica et biophysica acta, 816(1), 77-82 (1985-06-11)
Biotin uptake by Escherichia coli K12 has been reinvestigated. The vitamin uptake is an active process depending on energy and inhibited by uncouplers. The kinetic parameters (Km = 0.27 microM, Vmax = 6.8 pmol/min per mg dry cells) are close
Dirk F H Winkler et al.
Proteomics, 8(5), 961-967 (2008-03-08)
Biotin-labeled peptides are used for numerous biochemical and microbiological applications. Due to the strong affinity of biotin to streptavidin, the detection of biotinylated molecules is very sensitive. A powerful technique for parallel synthesis and high-throughput screening of peptides is the
Biotin-containing reagents.
M Wilchek et al.
Methods in enzymology, 184, 123-138 (1990-01-01)
B T Miller et al.
Peptides, 18(10), 1585-1595 (1997-01-01)
N-hydroxysuccinimide (NHS) esters of biotin are reported to react specifically with amino groups of peptides and proteins. However, we have found that these reagents can readily acylate other functional groups in specific peptide sequences under relatively mild conditions. We have
D L Vesely
Science (New York, N.Y.), 216(4552), 1329-1330 (1982-06-18)
Biotin and its analog, (+)-biotin-p-nitrophenyl ester enhanced guanylate cyclase activity two- to threefold in rat liver, kidney, colon, cerebellum, and heart. Dose-response relationships revealed that at concentrations as low as 1 micromolar, both biotin and its analog caused maximal augmentation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service