Skip to Content
Merck
All Photos(1)

Key Documents

28918

Sigma-Aldrich

Cyclohexane

for luminescence, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

grade

for luminescence

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

Assay

≥99.5% (GC)
≥99.5%

autoignition temp.

500 °F

expl. lim.

9 %

refractive index

n20/D 1.426 (lit.)
n20/D 1.426

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)

fluorescence

λex 200-600 nm; λem 300-800 nm (corresponds)

λ

1 cm path, H2O reference

UV absorption

λ: 210 nm Amax: 0.7
λ: 220 nm Amax: 0.32
λ: 230 nm Amax: 0.11
λ: 240 nm Amax: 0.04
λ: 250 nm Amax: 0.008
λ: 260 nm Amax: 0.003

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trevor James Morgan et al.
PloS one, 10(8), e0136511-e0136511 (2015-08-27)
A reactor was designed and commissioned to study the fast pyrolysis behavior of banagrass as a function of temperature and volatiles residence time. Four temperatures between 400 and 600°C were examined as well as four residence times between ~1.0 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service