Skip to Content
Merck
All Photos(1)

Key Documents

U1757

Millipore

Urea agar base (Christensen)

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606

shelf life

limited shelf life, expiry date on the label

Quality Level

final pH

6.8±0.2 (25 °C)

application(s)

environmental
food and beverages
veterinary

microbiology

suitability

nonselective and differential for Proteus spp.

Application

Recommended for the detection of urease production.

Components

Ingredients (g/L)
Peptic digest of animal tissue, 1.00
Dextrose, 1.00
Sodium chloride, 5.00
Disodium phosphate, 1.20
Monopotassium phosphate, 0.80
Phenol red, 0.012
Agar, 15.00

Preparation Note

Supplement: 50 ml/l of sterile 40% Urea Solution (08582)

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MacFaddin,J.
Biochemical Tests for Identification of Medical Bacteria (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service