Skip to Content
Merck
All Photos(2)

Documents

NIST25D

Manganese ore

NIST® SRM® 25d

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 60 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for use in validation of chemical and instrumental methods of analysis. A unit of SRM 25d consists of a bottle containing approximately 60 g of powder of particle size less than 74 μm (No. 200 sieve). For more information, please refer to the COA and SDS.

SRM 25D_cert

SRM 25D_SDS

Other Notes

Certified for the analytes listed below.
Manganese (Mn), Phosphorus pentoxide (P2O5), Silicon dioxide (SiO2)

Reference/Informational value is provided for the following analytes.
Aluminium oxide (Al2O3), Iron oxide (Fe2O3), Potassium oxide (K2O), Titanium dioxide (TiO2), Available Oxygen (O), Barium (Ba), Calcium oxide (CaO), Moisture

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service