Key Documents
GX208094053
Supelco
Scott Spray Chamber for SpectroBlue or Arcos II
About This Item
UNSPSC Code:
12352200
manufacturer/tradename
20-809-4053
compatibility
Spectro/Ametek Spectro Blue EOP
Spectro/Ametek Spectro Blue SOP
Choose from one of the most recent versions:
Certificates of Analysis (COA)