Skip to Content
Merck
All Photos(1)

Key Documents

DLW354210

Sigma-Aldrich

Corning® Poly-D-Lysine 20 mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

synthetic

Assay

>98%

form

lyophilized (from aqueous solution)

mol wt

500-550 kDa

packaging

pkg of 20 mg

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

Legal Information

Corning is a registered trademark of Corning, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service