Skip to Content
Merck
All Photos(1)

Key Documents

96415

Sigma-Aldrich

Cinnamonitrile

purum, ≥97.0% (GC)

Synonym(s):

3-Phenylacrylonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCN
CAS Number:
Molecular Weight:
129.16
Beilstein:
1209546
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.601 (lit.)
n20/D 1.601

bp

254-255 °C (lit.)

mp

18-20 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

SMILES string

N#C\C=C\c1ccccc1

InChI

1S/C9H7N/c10-8-4-7-9-5-2-1-3-6-9/h1-7H/b7-4+

InChI key

ZWKNLRXFUTWSOY-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service