Skip to Content
Merck
All Photos(1)

Documents

92607

Supelco

Malvidin chloride

analytical standard

Synonym(s):

3,4′,5,7-Tetrahydroxy-3′,5′-dimethoxyflavylium chloride, Malvidin, Oenidin chloride, Primulidin chloride, Syringidin chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClO7
CAS Number:
Molecular Weight:
366.75
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤8.0% water

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Cl-].COc1cc(cc(OC)c1O)-c2[o+]c3cc(O)cc(O)c3cc2O

InChI

1S/C17H14O7.ClH/c1-22-14-3-8(4-15(23-2)16(14)21)17-12(20)7-10-11(19)5-9(18)6-13(10)24-17;/h3-7H,1-2H3,(H3-,18,19,20,21);1H

InChI key

KQIKOUUKQBTQBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Anthocyanidin. Antioxidant.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chul Han An et al.
Plant physiology and biochemistry : PPB, 89, 1-10 (2015-02-15)
We previously reported that the transient and stable expression of IbMYB1a produced anthocyanin pigmentation in tobacco leaves and transgenic Arabidopsis plants, respectively. To further determine the effects of different promoters on the expression of IbMYB1a and anthocyanin production, we generated
Liga Saulite et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 53, 86-95 (2019-01-23)
Anthocyanidins are plant phytochemicals found at high concentrations in berries, vegetables and flowers. Anthocyanidins have been extensively investigated due to their antioxidative, antidiabetic and anti-inflammatory effects. Few studies show that anthocyanidins decrease obesity and improve bone density. However, the effects
Chloé Roullier-Gall et al.
PloS one, 9(5), e97615-e97615 (2014-05-27)
The chemical composition of grape berries is influenced by various environmental conditions often considered to be representative of a "terroir". If grapes from a given terroir are assumed to reflect this origin in their chemical compositions, the corresponding wine should
Claudio Inostroza-Blancheteau et al.
Plant physiology and biochemistry : PPB, 85, 85-95 (2014-11-15)
The effects of increased doses of UV-B radiation on anatomical, biochemical and molecular features of leaves of two highbush blueberry (Vaccinium corymbosum L. cv. Brigitta and Bluegold) genotypes were investigated. Plants were grown in a solid substrate and exposed to
Joseph A Lutz et al.
Fitoterapia, 98, 11-21 (2014-06-28)
The alpha7 nicotinic acetylcholine receptor (nAChR) is a potential target in neuroinflammation. Screening a plant extract library identified Solidago nemoralis as containing methyl-quercetin derivatives that are relatively selective ligands for the alpha7 nAChR. Flavonoids are not known for this activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service