Skip to Content
Merck
All Photos(1)

Key Documents

92491

Supelco

4-Methoxyphenylacetic acid

analytical standard

Synonym(s):

Homoanisic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CO2H
CAS Number:
Molecular Weight:
166.17
Beilstein:
1101737
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

bp

140 °C/3 mmHg (lit.)

mp

84-86 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(CC(O)=O)cc1

InChI

1S/C9H10O3/c1-12-8-4-2-7(3-5-8)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)

InChI key

NRPFNQUDKRYCNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methoxyphenylacetic acid belongs to the class of aryl alkanoic acids, with a broad-spectrum of applications, which includes its role as an intermediate in synthetic organic chemistry, building block material for the production of a variety of essential drugs, an additive in the manufacture of unsubstituted indigo dyes and corrosion inhibitor for aluminum in acidic and alkaline solutions. It is a structural analog of 2-(4-methoxyphenoxy)propanoic acid, reported to occur in cocoa products.

Application

4-Methoxyphenylacetic acid may be used as an analytical reference standard for the determination of the analyte in Illicium verum Hook. f. using gas chromatography coupled to mass spectrometry (GC-MS) technique and urine samples using high-performance liquid chromatography (HPLC) with ultraviolet and fluorescence detectors.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

379.4 °F

Flash Point(C)

193 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occurrence of 2-(4-methoxyphenoxy) propanoic acid in roasted coffee beans: analysis by gas-liquid chromatography and by high-performance liquid chromatography
Rathbone BE, et al.
Journal of Agricultural and Food Chemistry, 37(1), 54-58 (1989)
Separation of urinary aromatic acids and measurement of vanilmandelic acid by high-performance liquid chromatography
Molnar I, et al.
Chromatographia, 11(5), 260-265 (1978)
Effect of three extraction methods on the volatile component of Illicium verum Hook. f. analyzed by GC-MS
Qin W, et al.
Wuhan university journal of natural sciences, 12(3), 529-553 (2007)
Solubility measurement, modeling, and thermodynamic functions for para-methoxyphenylacetic acid in pure and mixed organic and aqueous systems
Tangirala R, et al.
Journal of Chemical and Engineering Data, 63(9), 3369-3338 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service