Skip to Content
Merck
All Photos(1)

Key Documents

90373

Supelco

2,5-Dimethyl-1H-pyrrole-3,4-dicarboxaldehyde

for HPLC derivatization, LiChropur, ≥98.5% (HPLC)

Synonym(s):

2,5-Dimethylpyrrole-3,4-dicarbaldehyde, 3,4-Diformyl-2,5-dimethylpyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
Beilstein:
118554
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for HPLC derivatization

Quality Level

Assay

≥98.5% (HPLC)

form

solid

quality

LiChropur

technique(s)

HPLC: suitable

SMILES string

CC1=C(C([H])=O)C(C([H])=O)=C(C)N1

InChI

1S/C8H9NO2/c1-5-7(3-10)8(4-11)6(2)9-5/h3-4,9H,1-2H3

InChI key

VJDZUXWJEWKBKE-UHFFFAOYSA-N

Application

suitable for derivatisation of amino acids

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service