Skip to Content
Merck
All Photos(1)

Documents

88122

Sigma-Aldrich

Tetrapropylammonium perchlorate

≥98.0% (T)

Synonym(s):

N,N,N-Tripropyl-1-propanaminium perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)4N(ClO4)
CAS Number:
Molecular Weight:
285.81
Beilstein:
3639544
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

reaction suitability

reagent type: oxidant

mp

238-242 °C

SMILES string

[O-]Cl(=O)(=O)=O.CCC[N+](CCC)(CCC)CCC

Looking for similar products? Visit Product Comparison Guide

Application

Investigations of:
  • The liquid junction potential between different solvents
  • The effect of cathode potential on electrochemical polymerization mechanism and physiochemical properties of styrene-Me methacrylate copolymers

Used in electrochemical:
  • Oxidation kinetics of ferrocene at platinum in MeCN containing tetraalkylammonium perchlorate using chronoamperometry or voltammetry
  • Reduction of methylbenzyl chloride on mercury
  • Impedance spectroscopy

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service