Skip to Content
Merck
All Photos(1)

Key Documents

80380065-0010

Dyed Functionalized Microspheres

K1-030 BLUE W-1G, polystyrene, carboxylic acid functionalized, 0.270-0.330 μm diameter, blue, vial of 1 g

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

material

polyethylene bottle
polystyrene

packaging

vial of 1 g

manufacturer/tradename

Estapor®

concentration

10% solids

color

blue

diameter

0.270-0.330 μm

capacity

200-500 μeq/g surface charge density

functional group

carboxylic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Estapor′s colored microspheres are best suited for assays where the primary goal is simply detecting the presence or absence of the target analyte. Choose the desired combination of surface functionalization, color and size to find the best microsphere for your application.

Application

Dyed microspheres are extremely uniform and are dyed internally (their surface properties are unchanged), insuring maximum color brilliance and preventing dye leaching in water or aqueous buffers.

Legal Information

Estapor is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service