Skip to Content
Merck
All Photos(1)

Documents

61792

Millipore

Leifson Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

DC Agar, Deoxycholate Citrate Agar according to Leifson, modified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
ammonium ferric citrate, 1 g/L
lactose, 10 g/L
meat extract, 5 g/L
meat peptone, 5 g/L
neutral red, 0.02 g/L
sodium citrate, 6 g/L
sodium deoxycholate, 3 g/L
sodium thiosulfate, 5.4 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.5±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages

microbiology

suitability

selective and differential for Salmonella spp.
selective and differential for Shigella spp.

Related Categories

Application

For the isolation of Salmonella and Shigella species according to Leifson, modified by Hynes.

Preparation Note

Dissolve 50.4 g in 1 litre distilled water heating if necessary, cool rapidly and pour plates. Do NOT autoclave.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E. Leifson
The Journal of Pathology and Bacteriology, 40, 581-581 (1935)
M.Hynes
The Journal of Pathology and Bacteriology, 54, 193-193 (1942)

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service