Skip to Content
Merck
All Photos(1)

Documents

53354

Supelco

1,3-Dimethoxybenzene

analytical standard

Synonym(s):

Dimethylresorcinol, Resorcinol dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
Beilstein:
878582
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

refractive index

n20/D 1.524 (lit.)

bp

85-87 °C/7 mmHg (lit.)

density

1.055 g/mL at 25 °C (lit.)

format

neat

SMILES string

COc1cccc(OC)c1

InChI

1S/C8H10O2/c1-9-7-4-3-5-8(6-7)10-2/h3-6H,1-2H3

InChI key

DPZNOMCNRMUKPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service