Skip to Content
Merck
All Photos(1)

Key Documents

45364

Sigma-Aldrich

1,2-Epoxyoctane

purum, ≥95.0% (GC)

Synonym(s):

1-Octene oxide, Hexyloxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O
CAS Number:
Molecular Weight:
128.21
Beilstein:
79912
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

purum

Assay

≥95.0% (GC)

refractive index

n20/D 1.420

bp

62-64 °C/17 mmHg (lit.)

density

0.835 g/mL at 20 °C (lit.)

application(s)

agriculture
environmental

storage temp.

2-8°C

SMILES string

CCCCCCC1CO1

InChI

1S/C8H16O/c1-2-3-4-5-6-8-7-9-8/h8H,2-7H2,1H3

InChI key

NJWSNNWLBMSXQR-UHFFFAOYSA-N

replaced by

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Levin et al.
Archives of biochemistry and biophysics, 220(2), 485-494 (1983-02-01)
Metabolism of cholesterol 5,6 alpha-oxide to the 5,6-glycol is catalyzed by a rat liver microsomal epoxide hydrolase that is distinct from the microsomal epoxide hydrolase that metabolizes a wide range of xenobiotic alkene and arene oxides. The two enzymes are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service