Skip to Content
Merck
All Photos(2)

Key Documents

37079

Supelco

Phenmedipham-ethyl

PESTANAL®, analytical standard

Synonym(s):

3-Ethoxycarbonylaminophenyl 3-methylcarbanilate, 3-[(Ethoxycarbonyl)amino]phenyl N-(3-methylphenyl)carbamate, Ethyl 3-(3-methylcarbaniloyloxy)carbanilate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N2O4
CAS Number:
Molecular Weight:
314.34
Beilstein:
2397745
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)Nc1cccc(OC(=O)Nc2cccc(C)c2)c1

InChI

1S/C17H18N2O4/c1-3-22-16(20)18-14-8-5-9-15(11-14)23-17(21)19-13-7-4-6-12(2)10-13/h4-11H,3H2,1-2H3,(H,18,20)(H,19,21)

InChI key

MVEFZZKZBYQFPP-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service