Skip to Content
Merck
All Photos(1)

Key Documents

34535

Supelco

3-Phenoxy-13C6-benzoic acid

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C6H5OC6H4CO2H
Molecular Weight:
220.17
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

SMILES string

O=C(O)C1=CC(O[13C]2=[13CH][13CH]=[13CH][13CH]=[13CH]2)=CC=C1

InChI

1S/C13H10O3/c14-13(15)10-5-4-8-12(9-10)16-11-6-2-1-3-7-11/h1-9H,(H,14,15)/i1+1,2+1,3+1,6+1,7+1,11+1

InChI key

NXTDJHZGHOFSQG-UJVDBDNBSA-N

General description

3-Phenoxy-13C6-benzoic acid is a 13C6 analog of phenoxybenzoic acid, a metabolite of pyrethroid insecticide, wherein the phenoxy carbons are substituted by 13C6.

Application

3-Phenoxy-13C6-benzoic acid may be used as an isotope-labeled pesticide standard to determine the levels of pyrethroid metabolites in human urine by liquid chromatography-tandem mass spectrometry (LC-MS/MS) and gas chromatography (GC) coupled to MS/MS.
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A method for the simultaneous quantification of eight metabolites of synthetic pyrethroids in urine of the general population using gas chromatography-tandem mass spectrometry
Schettgen T, et al.
Analytical and Bioanalytical Chemistry, 408(20), 5467-5478 (2016)
Simultaneous determination of five synthetic pyrethroid metabolites in urine by liquid chromatography-tandem mass spectrometry: application to 39 persons without known exposure to pyrethroids
Le Grand R, et al.
Toxicology Letters, 210(2), 248-253 (2012)
Microbial degradation of pesticide residues and an emphasis on the degradation of cypermethrin and 3-phenoxy benzoic acid: a review
Huang Y, et al.
Molecules (Basel), 23(9), 2313-2313 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service