Skip to Content
Merck
All Photos(1)

Key Documents

228699

Sigma-Aldrich

Sodium metaarsenite

98%

Synonym(s):

Sodium (meta)arsenite, Sodium dioxoarsenate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaAsO2
CAS Number:
Molecular Weight:
129.91
EC Number:
MDL number:
UNSPSC Code:
12352300

Assay

98%

density

1.87 g/mL at 25 °C (lit.)

SMILES string

[Na+].[O-][As]=O

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Xu et al.
Archives of toxicology, 89(7), 1071-1082 (2014-06-11)
Intercellular communications within the cancer microenvironment coordinate the assembly of various cell types. Exosomes are mediators of intercellular communication in immune signaling, tumor promotion, stress responses, and angiogenesis. The present research aimed to determine whether miRNAs secreted from human bronchial
Mirjam M Schaap et al.
Archives of toxicology, 89(12), 2413-2427 (2014-10-02)
Alternative methods to detect non-genotoxic carcinogens are urgently needed, as this class of carcinogens goes undetected in the current testing strategy for carcinogenicity under REACH. A complicating factor is that non-genotoxic carcinogens act through several distinctive modes of action, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service