Skip to Content
Merck
All Photos(1)

Documents

05981501

Lemon oil

secondary reference standard

Synonym(s):

Lemon, extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

matrix material

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service