Skip to Content
Merck
All Photos(1)

Key Documents

05334

Sigma-Aldrich

7-Aminoindole

≥95.0% (HPLC)

Synonym(s):

(Indol-7-yl)amine, 7-Indolylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (HPLC)

mp

96-100 °C

SMILES string

Nc1cccc2cc[nH]c12

InChI

1S/C8H8N2/c9-7-3-1-2-6-4-5-10-8(6)7/h1-5,10H,9H2

InChI key

WTFWZOSMUGZKNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Fluorescent anion receptors and sensors
  • Sulfate receptors
  • Protein Kinase C θ (PKCθ) inhibitors
  • Factor Xa Inhibitors
  • Aurora kinase inhibitor
  • Antagonist of Mineralocorticoid Receptor
  • Potent histone deacetylase (HDAC) inhibitors
  • TRPV1 antagonists
  • Derivatives of N-(7-indolyl)benzenesulfonamide as cell cycle inhibitors

Caution

may discolor to brown on storage

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service