Skip to Content
Merck
All Photos(1)

Documents

23-0550

Sigma-Aldrich

Oil of turpentine

SAJ first grade

Synonym(s):

Fir oil, Oil of turpentine, Pine oil, Turpentine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

SAJ first grade

vapor density

4.84 (−7 °C, vs air)

vapor pressure

4 mmHg ( −6.7 °C)

form

liquid

availability

available only in Japan

bp

153-175 °C (lit.)

mp

−55 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

InChI key

DCXXKSXLKWAZNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

'Myiasis-in-ano'.
Sijo K John et al.
Tropical gastroenterology : official journal of the Digestive Diseases Foundation, 33(3), 231-232 (2013-04-23)
Abitha Sukumaran et al.
Blood cells, molecules & diseases, 49(1), 11-19 (2012-04-17)
Anemia of inflammation is characterized by disturbances in systemic iron homeostasis. In order to better understand the events involved, we carried out a time-course study on the effects of acute and chronic inflammation on iron-related proteins in mouse splenic macrophages
Rahul Srivastava et al.
Indian journal of dental research : official publication of Indian Society for Dental Research, 22(4), 615-615 (2011-11-30)
Myiasis, a term first introduced by Hope (1840), refers to the invasion of tissues and organs of animals and human wounds and certain body cavities by the dipteran larvae which manifests as subcutaneous furunculoid or boillike lesions. Oral myiasis is
Mirela-Onita Iovu et al.
Carbohydrate research, 355, 63-68 (2012-05-29)
This study aimed to assess whether chronic administration of chondroitin sulfate (CS) affects baseline expression of cytochrome P450 isoforms and impedes the decrease in expression and activity of CYP1A2 and CYP3A6 in rabbits with a turpentine-induced inflammatory reaction (TIIR). Seven
Takashi Kuribayashi et al.
Experimental animals, 61(2), 125-130 (2012-04-26)
The aim of this study was to investigate the synthesis of α(2)-macroglobulin (α2M) in hepatopathic rats injected with turpentine oil to induce acute inflammation. Hepatopathy was induced by oral administration of acetaminophen at a dose of 1 g/kg daily for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service