Skip to Content
Merck
All Photos(1)

Documents

ZFCBIXXSP

Milli-Q

Country Box Milli-Q® IX Systems

Available only in Spain and Chile

Synonym(s):

Country Box Milli-Q® IX SP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104214
NACRES:
SB.51

packaging

pkg of 1 kit

availability

available only in (Spain and Chile)

compatibility

for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015

Looking for similar products? Visit Product Comparison Guide

General description

The Country Box for Milli-Q® IX 7003/05/10/15 pure water systems is an installation accessory that contains a printed Quick Reference Guide, a user manual in local language and in English on a USB key, and a power cord adapted to the local electrical network.

Application

The components of the country box assist users to quickly access the information they need to correctly use their system and connect to the local power network.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service