XX6700024
Vent/Relief Valve, 9 bar
Accessories for filter holders for sample preparation.
Synonym(s):
9 Bar Pressure Relief Valve, Pressure Relief Valve
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Quality Level
parameter
8.6 bar max. inlet pressure (125 psi)
I.D.
6.3 mm (0.25 in.)
compatibility
for use with 293-mm Stainless Steel Sanitary Filter Holder
for use with PL-1 Cartridge Housings
for use with PS-1 Cartridge Housings
shipped in
ambient
General description
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.
Tri-Clamp (TC) fittings are referred to by convention as 1 1/2", 2", etc. 1 1/2" TC fittings connect to 1" or 1 1/2" O.D. tube sizes. 2" TC fittings connect to 2" O.D. tube size only.
Tri-Clamp (TC) fittings are referred to by convention as 1 1/2", 2", etc. 1 1/2" TC fittings connect to 1" or 1 1/2" O.D. tube sizes. 2" TC fittings connect to 2" O.D. tube size only.
Application
Accessories for filter holders for sample preparation.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service