Skip to Content
Merck
All Photos(1)

Documents

MX-PLATE

Millipore

96-well Filter Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NA.84

sterility

non-sterile

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

plate size

96 wells

detection method

fluorometric

Application

Filter plate in a 96-well plate format.
For use with MILLIPLEX MAP assays, see MILLIPLEX MAP kit protocol for specific use instructions

Physical form

Color Code:Not Applicable

Other Notes

Automation Compatible:No

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service