Skip to Content
Merck
All Photos(1)

Key Documents

MABT406

Sigma-Aldrich

Anti-GPVI Antibody, clone 8E9, Capture Antibody

clone 8E9, from rat

Synonym(s):

Platelet glycoprotein VI, GPVI, Glycoprotein 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8E9, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GP6(51206)

General description

Glycoprotein VI (GPVI), or glycoprotein 6 (GP6), is a member of the immunoglobulin gene superfamily, closely related to Fc receptor gamma chain (FcRgamma) and natural killer receptors. GPVI is a collagen receptor involved in collagen-induced platelet adhesion and activation, and may contribute to arterial and venous thrombus formation. GPVI is a membrane glycoprotein that plays a crucial role in platelet procoagulant activity and subsequent thrombin and fibrin formation. GPVI is expressed in megakaryocytes and platelets. The signaling pathway of GPVI involves the FcRgamma, the Src kinases (likely Fyn/Lyn), the adapter protein LAT and leads to the activation of phospholipase C gamma2. It has been shown that bleeding disorders such as Bleeding Disorder Platelet-Type 11 (BDPLT11), a mild to moderate bleeding disorder caused by defective platelet activation and aggregation in response to collagen, is caused by defects in GPVI.

Immunogen

Recombinant protein corresponding to human GPVI.

Application

Anti-GPVI Antibody, clone 8E9, Capture Antibody is an antibody against GPVI for use in western blotting & ELISA.
ELISA Assay: A representative lot from an independent laboratory captured recombinant GPVI in a bead-based sandwich immunoassay (Bigalke, B., et al. (2011). Clin Chem. 57(6):898-904.). This antibody should be used in conjunction with Cat. No. MABT418, Anti-GPVI, clone 8A2, Detection Antibody, as an ELISA capture and detection antibody pair.

Quality

Evaluated by Western Blotting in recombinant human GPV1 glycoprotein.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected 0.1 µg of recombinant human GPV1 glycoprotein.

Target description

~50 kDa observed. The calculated molecular weight of this protein is 37 kDa However, a recombinant protein was observed at 50-55 kDa in Western Blotting.

Linkage

Replaces: MABT473

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service