Skip to Content
Merck
All Photos(3)

Key Documents

MABS1990M

Sigma-Aldrich

Anti-RhoB Antibody, clone 7F7

clone 7F7, from mouse

Synonym(s):

Rho-related GTP-binding protein RhoB, Rho cDNA clone 6, h6

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€128.00

€128.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 μL
€128.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€128.00


In StockDetails


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F7, monoclonal

species reactivity

rat, mouse, hamster, porcine, human, chicken, monkey, rabbit

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... RHOB(388)

General description

Rho-related GTP-binding protein RhoB (UniProt: P62745; also known as Rho cDNA clone 6, h6) is encoded by the RHOB (also known as ARH6, ARHB) gene (Gene ID: 388) in human. RhoB is a member of the Ras super-family of small GTP-binding proteins, which is involved in hydrolysis of GTP and is active in GTP-bound form. RhoB is a short-lived Rho GTPase whose expression is inducible by a variety of stimuli including growth factors and stress stimuli. Its levels are up-regulated by DNA damaging agents, such as hydrogen peroxide or ionizing radiation. RhoB plays a negative role in tumorigenesis as its deletion is known to cause tumor formation. It is reported to mediate apoptosis in neoplastically transformed cells following DNA damage. It is not considered to be essential for development, but affects cell adhesion and growth factor signaling in transformed cells. RhoB displays 83% homology with RhoA. However, unlike RhoA, which is cytosolic and translocates to plasma membrane upon activation, RhoB localizes to endosomes/multivesicular bodies. RhoB participates in sorting and degradation of growth factors and cytokine receptors. RhoB has three nucleotide-binding regions (aa 12-19; 59-63; and 117-120). RhoB is shown to be critically required for the inflammatory response of endothelial cells to TNF alpha, possibly through MAP kinase activation downstream of the TNFR.

Specificity

Clone 7F7 detects RhoB in multiple species. It targets an epitope within 19 amino acids from the C-terminal half. This clone specifically detects RhoB, but not RhoA, Rac1, or cdc42.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal half of human RhoB.

Application

Anti-RhoB, clone 7F7, Cat. No. MABS1990, is a mouse monoclonal antibody that detects Rho-related GTP-binding protein RhoB in human and has been tested for use in ELISA and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A 1:2,000 dilution from a representative lotdetected RhoB in 10 µg of MCF7 cell lysate.

Western Blotting Analysis: A representative lot detected RhoB in mouse and rabbit spleen, mouse kidney, recombinant RhoB, 293 cells expressing RhoB, CHO, H9C2, PK1P.185, COS, MDA MB-231, MCF7 and chicken fibroblasts (Courtesy of Lisa Laury-Kleintop, Ph.D., Lankenau Institute for Medical Research, Philadelphia PA, USA).

ELISA Analysis: A representative lot detected RhoB in Plate bound recombinant RhoB (Courtesy of Lisa Laury-Kleintop, Ph.D., Lankenau Institute for Medical Research, Philadelphia PA, USA).


Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected RhoB in 10 µg of HEK293 cell lysate.

Target description

~20 kDa observed; 22.12 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service